Assay Details
| Gene Symbol: B4galt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019804.4 | NP_062778.2 | 1 - 2 | 108 | 104 | |
| GenBank mRNA | AF142672.1 | - | 1 - 2 | 85 | 104 | |
| AK018658.1 | - | 1 - 2 | 72 | 104 | ||
| AK045602.1 | - | 1 - 2 | 80 | 104 | ||
| AK050005.1 | - | 1 - 2 | 104 | 104 | ||
| AK052013.1 | - | 1 - 2 | 102 | 104 | ||
| AK085368.1 | - | 1 - 2 | 94 | 104 | ||
| BC013492.1 | - | 1 - 2 | 71 | 4218187 | 104 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF142672.1,AK045602.1,AK050005.1,AK052013.1,AK085368.1,BC013492.1,NM_019804.4 transcripts.
Set Membership: |
|

B4galt4
Genomic Map