Assay Details
| Gene Symbol: Prmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019830.3 | NP_062804.1 | 2 - 3 | 189 | 80 | |
| GenBank mRNA | AF232716.1 | - | 2 - 3 | 101 | 80 | |
| BU524421.1 | - | 2 - 3 | 189 | 80 | ||
More Information
Set Membership: |
|

Prmt1
Genomic Map