Assay Details
Gene Symbol: Prl2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019991.1 | NP_064375.1 | 1 - 2 | 80 | 73 | |
GenBank mRNA | AB032428.1 | - | 1 - 2 | 101 | 73 | |
AF234636.1 | - | 1 - 2 | 80 | 73 | ||
AK005424.1 | - | 1 - 2 | 57 | 73 | ||
BC099458.1 | - | 1 - 2 | 76 | 30790018 | 73 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AB032428.1,AF234636.1,AK005424.1,BC099458.1,NM_019991.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P145511 | AF272368.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P145511 | AK005424.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P145511 | BC099458.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P145511 | NM_019991.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P145511 | AB032428.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P145511 | AF272368.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P145511 | AK005424.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P145511 | BC099458.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P145511 | NM_019991.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P145511 | AB032428.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 631293 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 678796 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 333591 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 929376 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 725495 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 109825 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 103034 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 114432 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 166412 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 30720 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 151447 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 4532 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P119305 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P150168 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P106921 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P145511 | AF272368.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P145511 | AK005424.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P145511 | BC099458.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P145511 | NM_019991.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P145511 | AB032428.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P145511 | AF272368.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P145511 | AK005424.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P145511 | BC099458.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P145511 | NM_019991.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P145511 | AB032428.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903597 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903602 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903618 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903603 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903630 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903627 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903599 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903607 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903626 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903606 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903598 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903634 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903631 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903610 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903616 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903613 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903601 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903614 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903612 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903624 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903622 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903623 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903628 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903635 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903632 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903608 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903605 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903620 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P106921 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P119305 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P150168 | - | - |
Whole Mouse Genome G4122A | A_51_P145511 | AF272368.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P145511 | AK005424.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P145511 | BC099458.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P145511 | NM_019991.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P145511 | AB032428.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P145511 | AF272368.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P145511 | AK005424.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P145511 | BC099458.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P145511 | NM_019991.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P145511 | AB032428.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134404 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P119305 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P150168 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P106921 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11008446 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11008453 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11008454 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10581995 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11008452 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903581 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903686 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903613 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903654 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903671 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903647 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903650 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903660 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903662 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903598 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903627 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903679 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903672 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903649 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903616 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903580 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903578 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903614 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903631 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903638 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903697 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903692 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903596 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903665 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903688 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903696 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903594 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903593 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903604 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903595 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903646 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903675 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903683 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903579 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903699 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903680 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903643 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903622 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903703 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903669 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903624 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903606 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903620 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903607 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903591 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903657 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903690 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903704 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903701 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903585 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903639 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903610 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903656 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903698 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903694 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903597 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903663 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903600 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903601 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903634 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903588 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903651 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903637 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903626 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903618 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903670 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903582 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903633 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903595 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903613 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903650 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903616 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903600 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11030994 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903620 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903594 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903618 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903603 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903607 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903651 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903610 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903643 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903626 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903649 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903622 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903638 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903596 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903654 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903646 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903657 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903666 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903663 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11031005 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903671 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903634 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10888944 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903637 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903624 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903602 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903623 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903627 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903669 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903593 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903665 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903662 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903670 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903606 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10742702 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903642 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903631 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903672 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903639 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903632 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903591 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903598 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903604 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903614 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903628 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903612 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903660 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28903656 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10404327 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10404326 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10404325 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10404329 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10404328 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10404324 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1449032_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10404323 | - | - |
Mouse Genome 430 2.0 Array | 1449032_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1449032_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56635_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56635_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1300001887.mm.1 | NM_019991.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300001887.mm.1 | NM_019991.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300001887.mm.1 | TC1300000304.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1300001888.mm.1 | NM_019991.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300001888.mm.1 | NM_019991.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300001888.mm.1 | TC1300000304.mm.1 | MTA TC |
Mouse Expression Set 430 | 1449032_PM_at | - | - |
Mouse Expression Set 430 | 1449032_at | - | - |
Mouse Genome 430A 2.0 Array | 1449032_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1449032_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1449032_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1449032_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 907747 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 3800500 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0003800500 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1227631 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1227631 | - | - |
mouse-6 v1.1 (Illumina) | scl44962.5.1_49-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 3800500 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl44962.5.1_49-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3800500 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1227631 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1227631 | - | - |
