| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0400020364.mm.1 |
TC0400002515.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400020364.mm.1 |
NM_020035.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400020367.mm.1 |
TC0400002515.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400020367.mm.1 |
NM_020035.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400020372.mm.1 |
TC0400002515.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400020372.mm.1 |
NM_020035.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400020366.mm.1 |
TC0400002515.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400020366.mm.1 |
NM_020035.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400020373.mm.1 |
TC0400002515.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400020373.mm.1 |
NM_020035.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400020365.mm.1 |
TC0400002515.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400020365.mm.1 |
NM_020035.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400020368.mm.1 |
TC0400002515.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400020368.mm.1 |
NM_020035.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400020370.mm.1 |
TC0400002515.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400020370.mm.1 |
NM_020035.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400020371.mm.1 |
TC0400002515.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400020371.mm.1 |
NM_020035.3 |
Refseq |