| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0900006287.mm.1 |
NM_020043.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900006287.mm.1 |
TC0900000829.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900006289.mm.1 |
NM_020043.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900006289.mm.1 |
TC0900000829.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900006285.mm.1 |
NM_020043.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900006285.mm.1 |
TC0900000829.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900006286.mm.1 |
NM_020043.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900006286.mm.1 |
TC0900000829.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900006288.mm.1 |
NM_020043.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900006288.mm.1 |
TC0900000829.mm.1 |
MTA TC |