Assay Details
| Gene Symbol: Igdcc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290315.1 | NP_001277244.1 | 8 - 9 | 1620 | 66 | |
| NM_020043.3 | NP_064427.2 | 8 - 9 | 1620 | 66 | ||
| GenBank mRNA | AB052620.1 | - | 8 - 9 | 1620 | 66 | |
| AB052621.1 | - | 8 - 9 | 1620 | 66 | ||
| AF176694.1 | - | 8 - 9 | 1571 | 66 | ||
| AK122535.1 | - | 1696 | 66 | |||
| CB518504.1 | - | 1 - 2 | 166 | 66 | ||
| CF535379.1 | - | 1 - 2 | 137 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Igdcc4
Genomic Map