Assay Details
| Gene Symbol: Igdcc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290315.1 | NP_001277244.1 | 10 - 11 | 1931 | 93 | |
| NM_020043.3 | NP_064427.2 | 10 - 11 | 1928 | 93 | ||
| GenBank mRNA | AB052620.1 | - | 10 - 11 | 1928 | 93 | |
| AB052621.1 | - | 10 - 11 | 1931 | 93 | ||
| AF176694.1 | - | 10 - 11 | 1879 | 93 | ||
| AK122535.1 | - | 2007 | 93 | |||
| AK143256.1 | - | 9 - 10 | 2390 | 93 | ||
| CB518504.1 | - | 3 - 4 | 474 | 93 | ||
| CF535379.1 | - | 3 - 4 | 448 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Igdcc4
Genomic Map