Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200002160.mm.1 |
NM_001039386.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002160.mm.1 |
NM_001039386.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002160.mm.1 |
NM_001039387.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002160.mm.1 |
NM_001039387.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002160.mm.1 |
NM_001177654.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002160.mm.1 |
NM_001177654.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002160.mm.1 |
NM_001177655.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002160.mm.1 |
NM_001177655.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002160.mm.1 |
NM_020276.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002160.mm.1 |
NM_020276.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002160.mm.1 |
TC0200000395.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200002158.mm.1 |
NM_001039386.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002158.mm.1 |
NM_001039386.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002158.mm.1 |
NM_001039387.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002158.mm.1 |
NM_001039387.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002158.mm.1 |
NM_001177654.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002158.mm.1 |
NM_001177654.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002158.mm.1 |
NM_001177655.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002158.mm.1 |
NM_001177655.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002158.mm.1 |
NM_020276.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002158.mm.1 |
NM_020276.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002158.mm.1 |
TC0200000395.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200002164.mm.1 |
NM_001039386.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002164.mm.1 |
NM_001039386.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002164.mm.1 |
NM_001039387.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002164.mm.1 |
NM_001039387.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002164.mm.1 |
NM_001177654.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002164.mm.1 |
NM_001177654.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002164.mm.1 |
NM_001177655.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002164.mm.1 |
NM_001177655.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002164.mm.1 |
NM_020276.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002164.mm.1 |
NM_020276.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002164.mm.1 |
TC0200000395.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200002165.mm.1 |
NM_001039386.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002165.mm.1 |
NM_001039386.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002165.mm.1 |
NM_001039387.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002165.mm.1 |
NM_001039387.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002165.mm.1 |
NM_001177654.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002165.mm.1 |
NM_001177654.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002165.mm.1 |
NM_001177655.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002165.mm.1 |
NM_001177655.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002165.mm.1 |
NM_020276.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002165.mm.1 |
NM_020276.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002165.mm.1 |
TC0200000395.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200002166.mm.1 |
NM_001039386.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002166.mm.1 |
NM_001039386.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002166.mm.1 |
NM_001039387.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002166.mm.1 |
NM_001039387.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002166.mm.1 |
NM_001177654.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002166.mm.1 |
NM_001177654.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002166.mm.1 |
NM_001177655.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002166.mm.1 |
NM_001177655.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002166.mm.1 |
NM_020276.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002166.mm.1 |
NM_020276.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002166.mm.1 |
TC0200000395.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200002156.mm.1 |
NM_001039386.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002156.mm.1 |
NM_001039386.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002156.mm.1 |
NM_001039387.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002156.mm.1 |
NM_001039387.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002156.mm.1 |
NM_001177654.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002156.mm.1 |
NM_001177654.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002156.mm.1 |
NM_001177655.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002156.mm.1 |
NM_001177655.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002156.mm.1 |
NM_020276.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002156.mm.1 |
NM_020276.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002156.mm.1 |
TC0200000395.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200002157.mm.1 |
NM_001039386.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002157.mm.1 |
NM_001039386.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002157.mm.1 |
NM_001039387.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002157.mm.1 |
NM_001039387.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002157.mm.1 |
NM_001177654.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002157.mm.1 |
NM_001177654.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002157.mm.1 |
NM_001177655.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002157.mm.1 |
NM_001177655.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002157.mm.1 |
NM_020276.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002157.mm.1 |
NM_020276.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002157.mm.1 |
TC0200000395.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200002162.mm.1 |
NM_001039386.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002162.mm.1 |
NM_001039386.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002162.mm.1 |
NM_001039387.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002162.mm.1 |
NM_001039387.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002162.mm.1 |
NM_001177654.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002162.mm.1 |
NM_001177654.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002162.mm.1 |
NM_001177655.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002162.mm.1 |
NM_001177655.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002162.mm.1 |
NM_020276.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002162.mm.1 |
NM_020276.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002162.mm.1 |
TC0200000395.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200002163.mm.1 |
NM_001039386.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002163.mm.1 |
NM_001039386.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002163.mm.1 |
NM_001039387.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002163.mm.1 |
NM_001039387.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002163.mm.1 |
NM_001177654.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002163.mm.1 |
NM_001177654.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002163.mm.1 |
NM_001177655.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002163.mm.1 |
NM_001177655.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002163.mm.1 |
NM_020276.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002163.mm.1 |
NM_020276.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200002163.mm.1 |
TC0200000395.mm.1 |
MTA TC |