Assay Details
Gene Symbol: B4galt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020579.2 | NP_065604.2 | 3 - 4 | 553 | 88 | |
GenBank mRNA | AF142671.1 | - | 3 - 4 | 493 | 88 | |
AK155471.1 | - | 4 - 5 | 653 | 88 | ||
BU703242.1 | - | 3 - 4 | 552 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
