Assay Details
| Gene Symbol: Fhl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021318.3 | NP_067293.1 | 4 - 5 | 691 | 91 | |
| GenBank mRNA | AF083394.1 | - | 4 - 5 | 639 | 91 | |
| AK006414.1 | - | 4 - 5 | 680 | 91 | ||
| BC048473.1 | - | 4 - 5 | 691 | 6770515 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fhl5
Genomic Map