Assay Details
| Gene Symbol: Cx3cl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB030188.1 | - | 1 | 263 | 140 | |
| BC057891.1 | - | 1 | 245 | 140 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Cx3cl1
Genomic Map