Assay Details
Gene Symbol: Ap4m1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021392.4 | NP_067367.3 | 11 - 12 | 1102 | 62 | |
GenBank mRNA | AF242858.1 | - | 11 - 12 | 970 | 62 | |
AK031638.1 | - | 11 - 12 | 1105 | 62 | ||
BC011174.1 | - | 11 - 12 | 987 | 4207033 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
