Assay Details
| Gene Symbol: Hs1bp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021429.3 | NP_067404.2 | 2 - 3 | 225 | 67 | |
| GenBank mRNA | AJ132192.1 | - | 2 - 3 | 219 | 67 | |
| AK040654.1 | - | 2 - 3 | 225 | 67 | ||
| AK084802.1 | - | 2 - 3 | 218 | 67 | ||
| AK154696.1 | - | 2 - 3 | 221 | 67 | ||
| AK170838.1 | - | 2 - 3 | 212 | 67 | ||
| BC095977.1 | - | 2 - 3 | 222 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hs1bp3
Genomic Map