Assay Details
Gene Symbol: Hs1bp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021429.3 | NP_067404.2 | 3 - 4 | 443 | 119 | |
GenBank mRNA | AJ132192.1 | - | 3 - 4 | 437 | 119 | |
AK040654.1 | - | 3 - 4 | 443 | 119 | ||
AK084802.1 | - | 3 - 4 | 436 | 119 | ||
AK154696.1 | - | 3 - 4 | 439 | 119 | ||
AK170838.1 | - | 3 - 4 | 430 | 119 | ||
AK185913.1 | - | 95 | 119 | |||
BC095977.1 | - | 3 - 4 | 440 | 119 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
