Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400009653.mm.1 |
NM_021434.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400009653.mm.1 |
NM_021434.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400009653.mm.1 |
TC1400001275.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400009649.mm.1 |
NM_021434.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400009649.mm.1 |
NM_021434.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400009649.mm.1 |
TC1400001275.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400009650.mm.1 |
NM_021434.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400009650.mm.1 |
NM_021434.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400009650.mm.1 |
TC1400001275.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400009651.mm.1 |
NM_021434.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400009651.mm.1 |
NM_021434.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400009651.mm.1 |
TC1400001275.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400009652.mm.1 |
NM_021434.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400009652.mm.1 |
NM_021434.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400009652.mm.1 |
TC1400001275.mm.1 |
MTA TC |