Assay Details
| Gene Symbol: Trappc2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021502.2 | NP_067477.1 | 2 - 3 | 239 | 82 | |
| GenBank mRNA | AB030200.1 | - | 2 - 3 | 232 | 82 | |
| AK007526.1 | - | 2 - 3 | 209 | 82 | ||
| BC028502.1 | - | 2 - 3 | 219 | 1366538 | 82 | |
| BC114968.1 | - | 207 | 82 | |||
| BY091059.1 | - | 2 - 3 | 240 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Trappc2l
Genomic Map