Assay Details
Gene Symbol: Sh3rf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021506.2 | NP_067481.2 | 7 - 8 | 1633 | 86 | |
GenBank mRNA | AF030131.1 | - | 7 - 8 | 1535 | 86 | |
AK045470.1 | - | 5 - 6 | 768 | 86 | ||
AK148137.1 | - | 7 - 8 | 1564 | 86 | ||
AK173185.1 | - | 7 - 8 | 1999 | 86 | ||
BC060113.1 | - | 8 - 9 | 1505 | 6405370 | 86 | |
BC060696.1 | - | 8 - 9 | 1505 | 6405370 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
