Assay Details
| Gene Symbol: Pnkp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290764.1 | NP_001277693.1 | 3 - 4 | 364 | 59 | |
| NM_001290766.1 | NP_001277695.1 | 2 - 3 | 456 | 59 | ||
| NM_001290767.1 | NP_001277696.1 | 2 - 3 | 199 | 59 | ||
| NM_021549.3 | NP_067524.2 | 2 - 3 | 456 | 59 | ||
| GenBank mRNA | AF129451.1 | - | 2 - 3 | 198 | 59 | |
| AK049992.1 | - | 2 - 3 | 259 | 59 | ||
| AK159535.1 | - | 2 - 3 | 222 | 59 | ||
| AK159770.1 | - | 2 - 3 | 222 | 59 | ||
| AK160020.1 | - | 2 - 3 | 222 | 59 | ||
| AK160160.1 | - | 3 - 4 | 360 | 59 | ||
| AK172558.1 | - | 2 - 3 | 59 | |||
| AK186762.1 | - | 2 - 3 | 69 | 59 | ||
| BC057659.1 | - | 2 - 3 | 258 | 2865792 | 59 | |
| BE627794.1 | - | 2 - 3 | 421 | 59 | ||
| CK628081.1 | - | 2 - 3 | 268 | 59 | ||
| CX223963.1 | - | 2 - 3 | 443 | 59 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001290767.1 transcripts.
Set Membership: |
|

Pnkp
Genomic Map