Assay Details
Gene Symbol: Nek7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021605.3 | NP_067618.1 | 8 - 9 | 859 | 60 | |
GenBank mRNA | AF217650.1 | - | 7 - 8 | 710 | 60 | |
AK035502.1 | - | 8 - 9 | 858 | 60 | ||
AK088173.1 | - | 8 - 9 | 908 | 60 | ||
AK149308.1 | - | 8 - 9 | 907 | 60 | ||
BC026541.1 | - | 8 - 9 | 849 | 60 | ||
BC037697.1 | - | 8 - 9 | 836 | 3994871 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
