Assay Details
| Gene Symbol: B4galt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022305.4 | NP_071641.1 | 2 - 3 | 1388 | 63 | |
| GenBank mRNA | AK029474.1 | - | 2 - 3 | 641 | 63 | |
| AK131721.1 | - | 2 - 3 | 491 | 63 | ||
| AK132970.1 | - | 2 - 3 | 1389 | 63 | ||
| AK154393.1 | - | 2 - 3 | 847 | 63 | ||
| AK167322.1 | - | 2 - 3 | 647 | 63 | ||
| BC053006.1 | - | 2 - 3 | 849 | 5698356 | 63 | |
| D00314.1 | - | 2 - 3 | 802 | 63 | ||
| J03880.1 | - | 2 - 3 | 827 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

B4galt1
Genomic Map