Assay Details
Gene Symbol: Ndst4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022565.2 | NP_072087.2 | 11 - 12 | 2945 | 65 | |
GenBank mRNA | AB036838.1 | - | 10 - 11 | 2395 | 65 | |
AK132938.1 | - | 6 - 7 | 1100 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
