Assay Details
| Gene Symbol: Polr1e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285800.1 | NP_001272729.1 | 11 - 12 | 1137 | 74 | |
| NM_022811.3 | NP_073722.1 | 9 - 10 | 993 | 74 | ||
| GenBank mRNA | AK050780.1 | - | 941 | 74 | ||
| AK083948.1 | - | 9 - 10 | 963 | 74 | ||
| AK132556.1 | - | 9 - 10 | 964 | 74 | ||
| BC034783.1 | - | 11 - 12 | 1084 | 4022372 | 74 | |
| D14336.1 | - | 9 - 10 | 956 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Polr1e
Genomic Map