Assay Details
| Gene Symbol: Polr1e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285800.1 | NP_001272729.1 | 12 - 13 | 1220 | 76 | |
| NM_022811.3 | NP_073722.1 | 10 - 11 | 1076 | 76 | ||
| GenBank mRNA | AK050780.1 | - | 1024 | 76 | ||
| AK083948.1 | - | 10 - 11 | 1046 | 76 | ||
| AK132556.1 | - | 10 - 11 | 1047 | 76 | ||
| BC034783.1 | - | 12 - 13 | 1167 | 4022372 | 76 | |
| D14336.1 | - | 10 - 11 | 1039 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Polr1e
Genomic Map