Assay Details
| Gene Symbol: Aspa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023113.5 | NP_075602.2 | 4 - 5 | 780 | 170 | |
| GenBank mRNA | AF212998.1 | - | 4 - 5 | 630 | 170 | |
| AK002859.1 | - | 4 - 5 | 778 | 170 | ||
| AK008354.1 | - | 4 - 5 | 783 | 170 | ||
| BC024934.1 | - | 4 - 5 | 744 | 4012726 | 170 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aspa
Genomic Map