Assay Details
Gene Symbol: Hspbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024172.3 | NP_077134.1 | 5 - 6 | 948 | 116 | |
GenBank mRNA | AF338351.1 | - | 4 - 5 | 886 | 116 | |
AK013263.1 | - | 5 - 6 | 939 | 116 | ||
AK088469.1 | - | 5 - 6 | 924 | 116 | ||
AK160128.1 | - | 5 - 6 | 944 | 116 | ||
BC014758.1 | - | 5 - 6 | 948 | 4014661 | 116 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
