Assay Details
| Gene Symbol: Mob4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025283.3 | NP_079559.2 | 4 - 5 | 321 | 87 | |
| GenBank mRNA | AK020446.1 | - | 4 - 5 | 277 | 87 | |
| AK147857.1 | - | 4 - 5 | 280 | 87 | ||
| AK162327.1 | - | 4 - 5 | 308 | 87 | ||
| BC037499.1 | - | 4 - 5 | 274 | 1512578 | 87 | |
| BC052475.1 | - | 4 - 5 | 266 | 87 | ||
| BC058168.1 | - | 4 - 5 | 274 | 4980300 | 87 | |
| U01138.1 | - | 4 - 5 | 265 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mob4
Genomic Map