Assay Details
| Gene Symbol: Sdhc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025321.3 | NP_079597.2 | 3 - 4 | 211 | 70 | |
| GenBank mRNA | AK002459.1 | - | 3 - 4 | 213 | 70 | |
| AK012818.1 | - | 3 - 4 | 212 | 70 | ||
| AK032458.1 | - | 213 | 70 | |||
| AK135503.1 | - | 3 - 4 | 213 | 70 | ||
| BC005779.1 | - | 3 - 4 | 198 | 2810852 | 70 | |
| BC083091.1 | - | 3 - 4 | 204 | 30445339 | 70 | |
| BY711932.1 | - | 3 - 4 | 212 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sdhc
Genomic Map