Assay Details
Gene Symbol: Zfp524 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025324.2 | NP_079600.1 | 1 - 2 | 58 | 97 | |
GenBank mRNA | AK002594.1 | - | 1 - 2 | 54 | 97 | |
AK011620.1 | - | 1 - 2 | 59 | 97 | ||
AK154328.1 | - | 1 - 2 | 65 | 97 | ||
BC019995.1 | - | 1 - 2 | 71 | 4458724 | 97 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK002594.1,AK011620.1,AK154328.1,BC019995.1,NM_025324.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
biological_process
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
nucleic acid binding
DNA binding
metal ion binding
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
biological_process
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
nucleic acid binding
DNA binding
metal ion binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24951574 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24951589 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24951576 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24951591 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24951577 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24951575 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24951592 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24951594 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P452798 | AK011620.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P452798 | AK154328.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P452798 | BC019995.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P452798 | NM_025324.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P452798 | AK011620.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P452798 | AK154328.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P452798 | BC019995.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P452798 | NM_025324.2 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 175719 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P144592 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P452798 | AK011620.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P452798 | AK154328.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P452798 | BC019995.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P452798 | NM_025324.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P452798 | AK011620.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P452798 | AK154328.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P452798 | BC019995.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P452798 | NM_025324.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P452798 | AK011620.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P452798 | AK154328.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P452798 | BC019995.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P452798 | NM_025324.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P452798 | AK011620.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P452798 | AK154328.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P452798 | BC019995.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P452798 | NM_025324.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24951595 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24951585 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24951582 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24951586 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24951590 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24951594 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24951575 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24951578 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24951592 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24951580 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24951596 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24951576 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24951588 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P144592 | - | - |
Whole Mouse Genome G4122A | A_51_P452798 | AK011620.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P452798 | AK154328.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P452798 | BC019995.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P452798 | NM_025324.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P452798 | AK011620.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P452798 | AK154328.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P452798 | BC019995.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P452798 | NM_025324.2 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P112838 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P128634 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P144592 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24951575 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24951594 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24951591 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24951592 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24951589 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24951577 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24951576 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24951574 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951616 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951531 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951622 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951520 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951599 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951525 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951529 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951601 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951590 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951651 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951573 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951588 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951623 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951580 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951602 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951607 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951549 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951603 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951586 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951543 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951636 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951596 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951656 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951536 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951644 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951641 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951548 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951513 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951539 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951554 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951592 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951619 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951639 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951629 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951518 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951578 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951628 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951652 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951521 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951550 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951533 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951519 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951560 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951640 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951571 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951643 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951538 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951621 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951527 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951575 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951542 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951594 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951557 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951544 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951653 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951582 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951609 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951569 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951535 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951615 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951617 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951614 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951611 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951515 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951555 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951624 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951627 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951585 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951598 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24951567 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24951594 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24951590 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24951582 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24951585 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24951583 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24951596 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24951588 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05997121 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06371035 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24951595 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24951578 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24951580 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24951592 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24951586 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10549749 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10549750 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1448915_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10549748 | - | - |
Mouse Genome 430 2.0 Array | 1448915_at | - | - |
Mouse Genome 430 2.0 Array | 1448915_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66056_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66056_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0700000620.mm.1 | TC0700000051.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700000620.mm.1 | NM_025324.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700000620.mm.1 | NM_025324.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700000619.mm.1 | TC0700000051.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700000619.mm.1 | NM_025324.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700000619.mm.1 | NM_025324.2 | Refseq |
Mouse Expression Set 430 | 1448915_at | - | - |
Mouse Expression Set 430 | 1448915_PM_at | - | - |
Murine Genome U74v2 Set | 103595_at | - | - |
Murine Genome U74v2 Set | 103595_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1448915_at | - | - |
Mouse Genome 430A 2.0 Array | 1448915_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1448915_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1448915_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 906752 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 430600 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 430600 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0000430600 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2438078 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2438078 | - | - |
mouse-6 v1.1 (Illumina) | scl33121.3_245-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 430600 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl33121.3_245-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 430600 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2438078 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2438078 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl33121.3_245-S | - | - |
