Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1300014219.mm.1 |
NM_025370.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300014219.mm.1 |
NM_025370.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300014219.mm.1 |
TC1300002201.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300014220.mm.1 |
NM_025370.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300014220.mm.1 |
NM_025370.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300014220.mm.1 |
TC1300002201.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300014221.mm.1 |
NM_025370.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300014221.mm.1 |
NM_025370.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300014221.mm.1 |
TC1300002201.mm.1 |
MTA TC |