Assay Details
| Gene Symbol: Nip7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025391.2 | NP_079667.2 | 4 - 5 | 478 | 82 | |
| NR_028367.1 | 3 - 4 | 571 | 82 | |||
| GenBank mRNA | AK003739.1 | - | 4 - 5 | 478 | 82 | |
| AK006057.1 | - | 4 - 5 | 458 | 82 | ||
| AK014290.1 | - | 4 - 5 | 437 | 82 | ||
| BC003972.1 | - | 4 - 5 | 465 | 3488726 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26142885 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26142883 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P411770 | BC068244.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P411770 | AK014290.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P411770 | AK003739.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P411770 | BC003972.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P411770 | NM_025391.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P411770 | BC068244.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P411770 | AK014290.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P411770 | AK003739.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P411770 | BC003972.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P411770 | NM_025391.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2253746 | AK018210.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2253746 | NM_025391.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2253746 | NR_028367.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2253746 | NM_001164472.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 234023 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P118435 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P411770 | BC068244.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P411770 | AK014290.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P411770 | AK003739.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P411770 | BC003972.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P411770 | NM_025391.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P411770 | BC068244.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P411770 | AK014290.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P411770 | AK003739.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P411770 | BC003972.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P411770 | NM_025391.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2253746 | AK018210.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2253746 | NM_025391.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2253746 | NR_028367.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2253746 | NM_001164472.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P411770 | BC068244.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P411770 | AK014290.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P411770 | AK003739.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P411770 | BC003972.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P411770 | NM_025391.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P411770 | BC068244.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P411770 | AK014290.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P411770 | AK003739.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P411770 | BC003972.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P411770 | NM_025391.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26142890 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26142887 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26142891 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26142884 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26142882 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26142883 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26142886 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26142885 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26142898 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P118435 | - | - |
| Whole Mouse Genome G4122A | A_51_P411770 | BC068244.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P411770 | AK014290.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P411770 | AK003739.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P411770 | BC003972.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P411770 | NM_025391.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P411770 | BC068244.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P411770 | AK014290.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P411770 | AK003739.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P411770 | BC003972.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P411770 | NM_025391.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P103176 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115160 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P118435 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06820053 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06820043 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06820052 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26142883 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26142885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26142920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26142925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26142915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26142882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26142907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26142880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26142884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26142936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26142885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26142891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26142912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26142906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26142938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26142914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26142923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26142886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26142929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26142934 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26142882 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26142891 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26142887 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26142899 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26142898 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26142889 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07220069 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26142885 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06946972 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26142886 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26142883 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10575147 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575148 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575145 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575149 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575146 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575150 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448480_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1453324_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10575144 | - | - |
| Mouse Genome 430 2.0 Array | 1448480_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1448480_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66164_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66164_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800010219.mm.1 | NM_001164472.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800010219.mm.1 | NM_001164472.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800010219.mm.1 | NM_025391.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800010219.mm.1 | NM_025391.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800010219.mm.1 | NR_028367.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800010219.mm.1 | NR_028367.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800010219.mm.1 | TC0800001263.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800010220.mm.1 | NM_001164472.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800010220.mm.1 | NM_001164472.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800010220.mm.1 | NM_025391.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800010220.mm.1 | NM_025391.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800010220.mm.1 | NR_028367.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800010220.mm.1 | NR_028367.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800010220.mm.1 | TC0800001263.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1448480_PM_at | - | - |
| Mouse Expression Set 430 | 1448480_at | - | - |
| Murine Genome U74v2 Set | 97305_at | - | - |
| Murine Genome U74v2 Set | 97305_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448480_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448480_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448480_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448480_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 330673 | - | - |
| ABI Mouse Genome Survey Microarray | 925122 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5890341 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4730039 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5890341 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6370286 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004730039 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006370286 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005890341 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2755730 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2718990 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2777151 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2755730 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2777151 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2718990 | - | - |
| mouse-6 v1.1 (Illumina) | scl066164.1_0-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0066164.2_158-S | - | - |
| mouse-6 v1.1 (Illumina) | scl000639.1_71-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6370286 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4730039 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5890341 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0066164.2_158-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl066164.1_0-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl000639.1_71-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4730039 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5890341 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6370286 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2755730 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2959122 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2718990 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2959122 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2755730 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2718990 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl066164.1_0-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl000639.1_71-S | - | - |

Nip7
Genomic Map