Assay Details
Gene Symbol: Med11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025397.2 | NP_079673.2 | 2 - 3 | 262 | 81 | |
GenBank mRNA | AK003984.1 | - | 2 - 3 | 240 | 81 | |
AK008153.1 | - | 2 - 3 | 280 | 81 | ||
BC025057.1 | - | 2 - 3 | 262 | 4238297 | 81 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27900777 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27900780 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27900779 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27900781 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27900782 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27900776 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27900778 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P349803 | BC025057.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P349803 | NM_025397.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P349803 | AK003984.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P349803 | BC025057.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P349803 | NM_025397.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P349803 | AK003984.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 600470 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P116408 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P349803 | BC025057.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P349803 | NM_025397.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P349803 | AK003984.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P349803 | BC025057.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P349803 | NM_025397.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P349803 | AK003984.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27900781 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27900795 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27900789 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27900771 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27900763 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27900772 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27900773 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27900761 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27900782 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27900783 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27900784 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27900756 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27900754 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27900759 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27900779 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27900757 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27900776 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27900775 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27900792 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27900791 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27900787 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27900767 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27900769 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27900764 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27900766 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27900786 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P116408 | - | - |
Whole Mouse Genome G4122A | A_51_P349803 | BC025057.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P349803 | NM_025397.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P349803 | AK003984.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P349803 | BC025057.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P349803 | NM_025397.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P349803 | AK003984.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P118898 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P116408 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09575340 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09575343 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09255967 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09133948 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09575335 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09119732 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09255965 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27900776 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27900779 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27900780 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27900782 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27900778 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27900777 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27900781 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900812 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900771 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900853 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900835 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900775 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900759 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900844 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900833 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900728 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900857 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900779 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900784 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900851 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900848 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900743 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900772 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900860 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900831 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900822 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900773 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900783 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900862 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900734 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900854 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900762 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900789 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900837 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900839 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900746 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900738 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900736 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900787 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900756 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900781 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900741 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900732 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900828 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900826 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900819 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900744 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900791 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900769 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900735 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900815 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900843 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900840 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900730 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900753 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900776 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27900818 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27900743 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27900795 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09042914 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09187440 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09042913 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09133948 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27900769 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27900779 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27900785 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27900744 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09255966 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27900781 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27900764 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27900746 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27900742 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09475676 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09475677 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09686418 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27900792 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27900789 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27900771 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27900776 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27900775 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27900787 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27900791 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27900757 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09739313 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09133947 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27900750 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09096887 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27900767 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27900759 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27900753 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27900783 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27900784 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27900756 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09169157 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10377824 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10377825 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10377823 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1417739_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10377822 | - | - |
Mouse Genome 430 2.0 Array | 1417739_at | - | - |
Mouse Genome 430 2.0 Array | 1417739_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66172_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66172_PM_at | - | - |
Mouse Expression Set 430 | 1417739_at | - | - |
Mouse Expression Set 430 | 1417739_PM_at | - | - |
Murine Genome U74v2 Set | 160596_at | - | - |
Murine Genome U74v2 Set | 160596_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1417739_at | - | - |
Mouse Genome 430A 2.0 Array | 1417739_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1417739_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1417739_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 471331 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 6350092 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006350092 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1257061 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1257061 | - | - |
mouse-6 v1.1 (Illumina) | scl41340.2.1_55-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6350092 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl41340.2.1_55-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6350092 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1257061 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_3000536 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3000536 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1257061 | - | - |
