Assay Details
Gene Symbol: Nat9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025400.3 | NP_079676.1 | 4 - 5 | 542 | 86 | |
GenBank mRNA | AK003948.1 | - | 4 - 5 | 496 | 86 | |
AK009262.1 | - | 3 - 4 | 272 | 86 | ||
AK082469.1 | - | 4 - 5 | 482 | 86 | ||
AK148048.1 | - | 4 - 5 | 574 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
molecular_function
dihydrolipoamide branched chain acyltransferase activity
sterol O-acyltransferase activity
N-acetyltransferase activity
O-acyltransferase activity
palmitoleoyl [acyl-carrier-protein]-dependent acyltransferase activity
carnitine O-acyltransferase activity
acetyltransferase activity
C-acyltransferase activity
palmitoyltransferase activity
N-acyltransferase activity
acylglycerol O-acyltransferase activity
serine O-acyltransferase activity
O-acetyltransferase activity
O-octanoyltransferase activity
octanoyltransferase activity
O-palmitoyltransferase activity
S-acyltransferase activity
S-acetyltransferase activity
S-malonyltransferase activity
malonyltransferase activity
C-palmitoyltransferase activity
transferase activity
transferase activity, transferring acyl groups
succinyltransferase activity
N-succinyltransferase activity
O-succinyltransferase activity
S-succinyltransferase activity
sinapoyltransferase activity
O-sinapoyltransferase activity
peptidyl-lysine N6-myristoyltransferase activity
peptidyl-lysine N6-palmitoyltransferase activity
benzoyl acetate-CoA thiolase activity
3-hydroxybutyryl-CoA thiolase activity
3-ketopimelyl-CoA thiolase activity
N-palmitoyltransferase activity
myristoyltransferase activity
acyl-CoA N-acyltransferase activity
protein-cysteine S-myristoyltransferase activity
protein-cysteine S-acyltransferase activity
dihydrolipoamide S-acyltransferase activity
glucosaminyl-phosphotidylinositol O-acyltransferase activity
ergosterol O-acyltransferase activity
lanosterol O-acyltransferase activity
naphthyl-2-oxomethyl-succinyl-CoA succinyl transferase activity
2,4,4-trimethyl-3-oxopentanoyl-CoA 2-C-propanoyl transferase activity
2-methylhexanoyl-CoA C-acetyltransferase activity
butyryl-CoA 2-C-propionyltransferase activity
2,6-dimethyl-5-methylene-3-oxo-heptanoyl-CoA C-acetyltransferase activity
L-2-aminoadipate N-acetyltransferase activity
UDP-3-O-[3-hydroxymyristoyl] glucosamine N-acyltransferase activity
keto acid formate lyase activity
Ras palmitoyltransferase activity
azetidine-2-carboxylic acid acetyltransferase activity
peptidyl-lysine N-acetyltransferase activity, acting on acetyl phosphate as donor
acetyl-CoA:L-lysine N6-acetyltransferase
biological_process
molecular_function
dihydrolipoamide branched chain acyltransferase activity
sterol O-acyltransferase activity
N-acetyltransferase activity
O-acyltransferase activity
palmitoleoyl [acyl-carrier-protein]-dependent acyltransferase activity
carnitine O-acyltransferase activity
acetyltransferase activity
C-acyltransferase activity
palmitoyltransferase activity
N-acyltransferase activity
acylglycerol O-acyltransferase activity
serine O-acyltransferase activity
O-acetyltransferase activity
O-octanoyltransferase activity
octanoyltransferase activity
O-palmitoyltransferase activity
S-acyltransferase activity
S-acetyltransferase activity
S-malonyltransferase activity
malonyltransferase activity
C-palmitoyltransferase activity
transferase activity
transferase activity, transferring acyl groups
succinyltransferase activity
N-succinyltransferase activity
O-succinyltransferase activity
S-succinyltransferase activity
sinapoyltransferase activity
O-sinapoyltransferase activity
peptidyl-lysine N6-myristoyltransferase activity
peptidyl-lysine N6-palmitoyltransferase activity
benzoyl acetate-CoA thiolase activity
3-hydroxybutyryl-CoA thiolase activity
3-ketopimelyl-CoA thiolase activity
N-palmitoyltransferase activity
myristoyltransferase activity
acyl-CoA N-acyltransferase activity
protein-cysteine S-myristoyltransferase activity
protein-cysteine S-acyltransferase activity
dihydrolipoamide S-acyltransferase activity
glucosaminyl-phosphotidylinositol O-acyltransferase activity
ergosterol O-acyltransferase activity
lanosterol O-acyltransferase activity
naphthyl-2-oxomethyl-succinyl-CoA succinyl transferase activity
2,4,4-trimethyl-3-oxopentanoyl-CoA 2-C-propanoyl transferase activity
2-methylhexanoyl-CoA C-acetyltransferase activity
butyryl-CoA 2-C-propionyltransferase activity
2,6-dimethyl-5-methylene-3-oxo-heptanoyl-CoA C-acetyltransferase activity
L-2-aminoadipate N-acetyltransferase activity
UDP-3-O-[3-hydroxymyristoyl] glucosamine N-acyltransferase activity
keto acid formate lyase activity
Ras palmitoyltransferase activity
azetidine-2-carboxylic acid acetyltransferase activity
peptidyl-lysine N-acetyltransferase activity, acting on acetyl phosphate as donor
acetyl-CoA:L-lysine N6-acetyltransferase
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28151867 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28151868 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2090415 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2167898 | AK148048.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2167898 | BC011315.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 153531 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 489065 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 550921 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2090415 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2167898 | AK148048.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2167898 | BC011315.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28151865 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28151860 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28151859 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28151840 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28151863 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28151836 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28151862 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28151868 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28151857 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28151842 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28151834 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28151847 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28151833 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28151831 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28151855 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28151850 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28151849 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28151841 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28151845 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28151864 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28151837 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28151830 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28151853 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28151843 | - | - |
Whole Mouse Genome G4122A | A_51_P467889 | AK082469.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P467889 | AK148048.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P467889 | AK154593.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P467889 | BC011315.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P467889 | NM_025400.3 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129554 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P104080 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09273461 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28151868 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28151867 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151765 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151840 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151867 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151860 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151862 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151836 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151834 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151853 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151864 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151830 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151863 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151837 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151772 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151768 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151842 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151761 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151843 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151857 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151847 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151850 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151770 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151769 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151859 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09506270 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28151862 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28151841 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28151842 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09506222 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28151860 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28151865 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28151867 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09741536 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28151831 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28151840 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09506247 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28151856 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28151854 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09273464 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28151847 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28151857 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09506224 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28151834 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28151853 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28151830 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28151859 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28151855 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28151843 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28151850 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10392861 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10392858 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10392857 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10392859 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10392860 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10392862 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1418781_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10392856 | - | - |
Mouse Genome 430 2.0 Array | 1418781_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1418781_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66176_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66176_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1100038099.mm.1 | NM_025400.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100038099.mm.1 | NM_025400.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100038099.mm.1 | TC1100004061.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100038098.mm.1 | NM_025400.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100038098.mm.1 | NM_025400.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100038098.mm.1 | TC1100004061.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100038097.mm.1 | NM_025400.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100038097.mm.1 | NM_025400.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100038097.mm.1 | TC1100004061.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100038101.mm.1 | NM_025400.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100038101.mm.1 | NM_025400.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100038101.mm.1 | TC1100004061.mm.1 | MTA TC |
Mouse Expression Set 430 | 1418781_at | - | - |
Mouse Expression Set 430 | 1418781_PM_at | - | - |
Murine Genome U74v2 Set | 168112_PM_r_at | - | - |
Murine Genome U74v2 Set | 168112_r_at | - | - |
Mouse Genome 430A 2.0 Array | 1418781_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1418781_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 910264 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 1410082 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 1410082 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0001410082 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2599145 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2599145 | - | - |
mouse-6 v1.1 (Illumina) | scl39347.7.18_70-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 1410082 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl39347.7.18_70-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1410082 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2599145 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2599145 | - | - |
