Assay Details
Gene Symbol: Cd302 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025422.4 | NP_079698.2 | 4 - 5 | 821 | 68 | |
GenBank mRNA | AK004267.1 | - | 4 - 5 | 757 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21257412 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21257457 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21257458 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21257459 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2075195 | AK004267.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2075195 | NM_025422.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2075195 | AK002812.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 454422 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 972169 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 11446 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 859804 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 955648 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 334788 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 658405 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 286606 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 172557 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 77913 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 849260 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 156789 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 314075 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 229202 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 134194 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 283951 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 384412 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 924557 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 818473 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 103011 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 98581 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 46639 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 141615 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P103476 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2075195 | AK004267.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2075195 | NM_025422.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2075195 | AK002812.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21257437 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21257469 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21257465 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21257474 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21257467 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21257478 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21257454 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21257441 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21257475 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21257459 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21257457 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21257456 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21257438 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21257443 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21257471 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21257463 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21257455 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21257452 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21257458 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21257480 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21257447 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21257479 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21257476 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21257451 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21257461 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21257449 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P103476 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00381005 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04535078 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00380996 | - | - |
Whole Mouse Genome G4122A | A_51_P514623 | AK004267.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106754 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P103476 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P00381005 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02107676 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01871226 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21257412 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21257458 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21257457 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21257459 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257253 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257415 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257390 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257465 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257238 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257343 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257479 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257469 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257363 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257416 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257260 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257351 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257476 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257380 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257348 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257367 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257251 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257346 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257248 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257355 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257478 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257443 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257236 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257474 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257378 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257252 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257259 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257451 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257433 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257308 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257376 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257429 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257368 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257258 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257485 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257265 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257391 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257441 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257342 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257449 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257304 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257400 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257431 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257438 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257352 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257501 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257337 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257454 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257464 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257410 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257330 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257322 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257267 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257398 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257339 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257482 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257306 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257395 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257375 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257264 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257321 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257426 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257334 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257239 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257503 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257370 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257320 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257471 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257256 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257345 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257295 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257235 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257365 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257247 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257502 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257392 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257261 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257459 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257244 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257437 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257301 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257388 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257319 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257412 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257249 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257435 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257421 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257311 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257299 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257241 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257413 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257254 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257424 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257315 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257245 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257349 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257233 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257371 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257263 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257324 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257377 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257313 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257310 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257495 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257475 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257491 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257456 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257402 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257297 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257243 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257461 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21257452 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21257388 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21257426 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21257419 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21257357 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21257433 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02015253 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21257437 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21257356 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21257429 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21257481 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02164158 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02015259 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02163088 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21257495 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21257454 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21257424 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21257502 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21257400 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21257435 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21257422 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21257415 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21257488 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02101968 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21257413 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21257431 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01788871 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21257498 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21257485 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21257478 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02191044 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21257361 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21257467 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01753026 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21257358 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21257441 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21257294 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21257501 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01507561 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01682779 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21257363 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21257482 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01285455 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21257406 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02077428 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21257503 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21257391 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21257410 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21257359 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21257456 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21257443 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21257416 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01753032 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21257461 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01270732 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21257465 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21257392 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21257449 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01753033 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02073793 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21257390 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01682777 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21257403 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01753025 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21257405 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21257398 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01815513 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21257418 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21257491 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21257447 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10482927 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10482923 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10482924 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10482922 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10482921 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10482925 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10482926 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10482928 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1448919_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1454374_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10482920 | - | - |
Mouse Genome 430 2.0 Array | 1454374_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1454374_at | - | - |
Mouse Genome 430 2.0 Array | 1448919_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1448919_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66205_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66205_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0200030664.mm.1 | NM_025422.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200030664.mm.1 | TC0200003599.mm.1 | MTA TC |
Mouse Expression Set 430 | 1448919_PM_at | - | - |
Mouse Expression Set 430 | 1448919_at | - | - |
Murine Genome U74v2 Set | 160387_at | - | - |
Murine Genome U74v2 Set | 160387_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1448919_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1448919_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1448919_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1448919_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1454374_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1454374_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 3850593 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 3850593 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0003850593 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2616097 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2616097 | - | - |
mouse-6 v1.1 (Illumina) | scl19239.6_43-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 3850593 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl19239.6_43-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3850593 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2616097 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2616097 | - | - |
