Assay Details
| Gene Symbol: Pno1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025443.2 | NP_079719.1 | 3 - 4 | 511 | 133 | |
| GenBank mRNA | AF349950.1 | - | 3 - 4 | 494 | 133 | |
| AK007324.1 | - | 3 - 4 | 476 | 133 | ||
| AK009146.1 | - | 3 - 4 | 478 | 133 | ||
| AK017153.1 | - | 3 - 4 | 482 | 133 | ||
| AK146152.1 | - | 3 - 4 | 505 | 133 | ||
| AK165942.1 | - | 3 - 4 | 472 | 133 | ||
| AK168177.1 | - | 3 - 4 | 505 | 133 | ||
| BC020037.1 | - | 3 - 4 | 494 | 4007940 | 133 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pno1
Genomic Map