Assay Details
| Gene Symbol: Pno1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025443.2 | NP_079719.1 | 5 - 6 | 693 | 90 | |
| GenBank mRNA | AF349950.1 | - | 5 - 6 | 676 | 90 | |
| AK007324.1 | - | 5 - 6 | 658 | 90 | ||
| AK009146.1 | - | 5 - 6 | 660 | 90 | ||
| AK017153.1 | - | 5 - 6 | 664 | 90 | ||
| AK146152.1 | - | 5 - 6 | 687 | 90 | ||
| AK165942.1 | - | 5 - 6 | 654 | 90 | ||
| AK168177.1 | - | 5 - 6 | 687 | 90 | ||
| BC020037.1 | - | 5 - 6 | 676 | 4007940 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27614822 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27614819 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27614817 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27614798 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27614823 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27614818 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27614820 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P192589 | AK168177.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P192589 | BC020037.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P192589 | AK146152.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P192589 | AK009896.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P192589 | AK165942.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P192589 | AK168177.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P192589 | BC020037.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P192589 | AK146152.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P192589 | AK009896.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P192589 | AK165942.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 727379 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 414504 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 124371 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 811618 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 84977 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P139965 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P150971 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P192589 | AK168177.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P192589 | BC020037.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P192589 | AK146152.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P192589 | AK009896.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P192589 | AK165942.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P192589 | AK168177.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P192589 | BC020037.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P192589 | AK146152.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P192589 | AK009896.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P192589 | AK165942.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27614810 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27614793 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27614807 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27614803 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27614792 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27614818 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27614794 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27614813 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27614820 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27614789 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27614800 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27614811 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27614823 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27614802 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27614788 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27614804 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27614815 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27614795 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27614808 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27614814 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27614801 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27614806 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27614799 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27614805 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P150971 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P139965 | - | - |
| Whole Mouse Genome G4122A | A_51_P192589 | AK168177.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P192589 | BC020037.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P192589 | AK146152.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P192589 | AK009896.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P192589 | AK165942.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P192589 | AK168177.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P192589 | BC020037.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P192589 | AK146152.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P192589 | AK009896.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P192589 | AK165942.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P119223 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114353 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P139965 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P150971 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08979014 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08979013 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09445318 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27614798 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27614822 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27614817 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27614823 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27614820 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27614818 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27614819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27614792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27614777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27614820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27614805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27614815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27614808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27614766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27614789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27614807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27614799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27614768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27614783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27614806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27614803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27614780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27614802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27614765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27614795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27614814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27614793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27614781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27614818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27614772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27614801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27614809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27614784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27614823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27614810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27614811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27614763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27614794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27614774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27614771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27614764 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27614810 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27614808 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27614807 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27614785 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27614778 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27614780 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27614787 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09245830 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27614815 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27614799 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27614784 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27614802 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27614794 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27614805 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27614781 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09245845 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27614783 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27614801 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27614820 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27614809 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27614813 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27614793 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27614792 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27614817 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27614814 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27614795 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09245837 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27614806 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10384475 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10374429 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10384482 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10384480 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10384477 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10374427 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10384479 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10374428 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10384478 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10384481 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10384476 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1450735_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10374426 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10384474 | - | - |
| Mouse Genome 430 2.0 Array | 1450735_at | - | - |
| Mouse Genome 430 2.0 Array | 1450735_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66249_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66249_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100021784.mm.1 | NM_025443.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100021784.mm.1 | NM_025443.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100021784.mm.1 | TC1100002251.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1450735_at | - | - |
| Mouse Expression Set 430 | 1450735_PM_at | - | - |
| Murine Genome U74v2 Set | 97237_PM_at | - | - |
| Murine Genome U74v2 Set | 97237_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450735_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450735_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450735_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450735_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 421496 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1090494 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1090494 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001090494 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1233889 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1233889 | - | - |
| mouse-6 v1.1 (Illumina) | scl40498.6_62-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1090494 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl40498.6_62-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1090494 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1233889 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1233889 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl40498.6_62-S | - | - |

Pno1
Genomic Map