Assay Details
| Gene Symbol: Tmem218 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025464.2 | NP_079740.1 | 3 - 4 | 282 | 65 | |
| GenBank mRNA | AK003010.1 | - | 3 - 4 | 279 | 65 | |
| AK007575.1 | - | 3 - 4 | 266 | 65 | ||
| AK008902.1 | - | 3 - 4 | 253 | 65 | ||
| AK161115.1 | - | 284 | 65 | |||
| BC034160.1 | - | 3 - 4 | 282 | 5049687 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P331207 | AK161115.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P331207 | BC034160.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P331207 | NM_025464.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P331207 | AK003010.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P331207 | AK007575.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P331207 | AK161115.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P331207 | BC034160.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P331207 | NM_025464.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P331207 | AK003010.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P331207 | AK007575.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 593808 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 344548 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 794474 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 284297 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 562521 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 801403 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 127398 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 933744 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 537173 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 573026 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 71219 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 169385 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P331207 | AK161115.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P331207 | BC034160.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P331207 | NM_025464.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P331207 | AK003010.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P331207 | AK007575.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P331207 | AK161115.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P331207 | BC034160.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P331207 | NM_025464.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P331207 | AK003010.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P331207 | AK007575.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26429386 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26429351 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26429363 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26429350 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26429344 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26429347 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26429387 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26429354 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26429364 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26429352 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26429377 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26429383 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26429343 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26429361 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26429370 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26429340 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26429372 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26429376 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26429374 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26429348 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26429355 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26429367 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26429366 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26429384 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26429371 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26429378 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26429358 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26429369 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26429349 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26429356 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26429380 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06752961 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06752940 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01906965 | - | - |
| Whole Mouse Genome G4122A | A_51_P331207 | AK161115.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P331207 | BC034160.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P331207 | NM_025464.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P331207 | AK003010.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P331207 | AK007575.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P331207 | AK161115.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P331207 | BC034160.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P331207 | NM_025464.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P331207 | AK003010.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P331207 | AK007575.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P126213 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P01906965 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07747191 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07747192 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07747203 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07747193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429386 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26429463 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07589569 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26429364 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07935996 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26429435 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26429399 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26429325 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08098134 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07534912 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26429372 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26429344 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26429363 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26429340 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07853196 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07747199 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26429385 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07714664 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26429358 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26429383 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07764966 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26429445 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07747194 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26429348 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26429393 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26429356 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26429404 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26429370 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26429369 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26429381 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26429350 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07839391 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07747200 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26429408 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26429402 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26429398 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26429448 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26429336 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26429380 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07543470 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26429443 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26429377 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26429359 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07908224 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26429371 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26429405 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26429450 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26429351 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26429374 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07865733 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26429347 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26429401 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26429440 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26429386 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26429389 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26429319 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26429442 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26429406 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26429454 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26429335 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26429451 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08162634 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26429453 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26429455 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26429396 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26429400 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26429438 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10584273 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10584275 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10584274 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10584272 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417174_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10584271 | - | - |
| Mouse Genome 430 2.0 Array | 1417174_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1417174_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66279_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66279_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900002639.mm.1 | NM_025464.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900002639.mm.1 | TC0900000358.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1417174_PM_at | - | - |
| Mouse Expression Set 430 | 1417174_at | - | - |
| Murine Genome U74v2 Set | 167693_PM_i_at | - | - |
| Murine Genome U74v2 Set | 167693_i_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417174_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417174_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417174_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417174_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 689299 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5340035 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5340035 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005340035 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2639402 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2639402 | - | - |
| mouse-6 v1.1 (Illumina) | scl37119.4.1_30-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5340035 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl37119.4.1_30-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5340035 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2639402 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2981896 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2981896 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2639402 | - | - |

Tmem218
Genomic Map