Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700003270.mm.1 |
TC0700000420.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700003270.mm.1 |
NM_025504.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700003270.mm.1 |
NM_025504.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700003281.mm.1 |
TC0700000420.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700003281.mm.1 |
NM_025504.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700003281.mm.1 |
NM_025504.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700003276.mm.1 |
TC0700000420.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700003276.mm.1 |
NM_025504.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700003276.mm.1 |
NM_025504.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700003279.mm.1 |
TC0700000420.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700003279.mm.1 |
NM_025504.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700003279.mm.1 |
NM_025504.4 |
Refseq |