Assay Details
| Gene Symbol: Atp5sl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290487.1 | NP_001277416.1 | 4 - 5 | 458 | 60 | |
| NM_025504.4 | NP_079780.2 | 4 - 5 | 1383 | 60 | ||
| GenBank mRNA | AK002370.1 | - | 4 - 5 | 438 | 60 | |
| AK009150.1 | - | 4 - 5 | 441 | 60 | ||
| AK039669.1 | - | 4 - 5 | 1384 | 60 | ||
| BC024867.1 | - | 3 - 4 | 414 | 4951552 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atp5sl
Genomic Map