Assay Details
| Gene Symbol: Atp5sl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290487.1 | NP_001277416.1 | 5 - 6 | 628 | 85 | |
| NM_025504.4 | NP_079780.2 | 5 - 6 | 1553 | 85 | ||
| GenBank mRNA | AK002370.1 | - | 5 - 6 | 608 | 85 | |
| AK009150.1 | - | 5 - 6 | 611 | 85 | ||
| AK039669.1 | - | 5 - 6 | 1554 | 85 | ||
| BC024867.1 | - | 4 - 5 | 584 | 4951552 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25010937 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2167000 | BC024867.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2167000 | NM_025504.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2167000 | AK002370.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 539629 | AK142395.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 539629 | AY195875.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 539629 | BC023090.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 539629 | BC027407.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 539629 | NM_153063.3 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 618610 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 581064 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 808076 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P142913 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2167000 | BC024867.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2167000 | NM_025504.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2167000 | AK002370.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010941 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010971 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010959 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010940 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010926 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010966 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010957 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010962 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010921 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010915 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010924 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010949 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010970 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010948 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010943 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010933 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010939 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010942 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010918 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010931 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010930 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010969 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010928 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010958 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010951 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010954 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010916 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010946 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010937 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010961 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010964 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010955 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010944 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010960 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25010922 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P142913 | - | - |
| Whole Mouse Genome G4122A | A_52_P330201 | AK021146.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P330201 | AK039669.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P330201 | NM_025504.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P402414 | AK009150.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P402414 | AK039669.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P402414 | BC024867.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P402414 | NM_025504.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P402414 | AK002370.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P118022 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P142913 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06120674 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06120671 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25010937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25011000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25011002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25011001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25010964 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010941 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010954 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06615458 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010948 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06565244 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010970 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010951 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010955 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010939 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010930 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010958 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010943 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010964 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010944 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010933 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010969 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010960 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010971 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06565245 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06419095 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010946 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010931 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010937 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010934 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010962 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010961 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010957 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010966 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06597145 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25010949 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10551167 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10551164 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10551163 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10551168 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10551165 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10551166 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416775_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1430450_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10551162 | - | - |
| Mouse Genome 430 2.0 Array | 1430450_at | - | - |
| Mouse Genome 430 2.0 Array | 1430450_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1416775_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1416775_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66349_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66349_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700003284.mm.1 | TC0700000420.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700003284.mm.1 | NM_025504.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700003284.mm.1 | NM_025504.4 | Refseq |
| Mouse Expression Set 430 | 1416775_PM_at | - | - |
| Mouse Expression Set 430 | 1416775_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416775_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416775_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416775_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416775_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430450_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430450_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 926124 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2370403 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002370403 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2666225 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2666225 | - | - |
| mouse-6 v1.1 (Illumina) | scl32925.7_312-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2370403 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl32925.7_312-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2370403 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2666225 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2666225 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl32925.7_312-S | - | - |

Atp5sl
Genomic Map