Assay Details
| Gene Symbol: Adprm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025510.3 | NP_079786.2 | 4 - 5 | 921 | 75 | |
| GenBank mRNA | AK009142.1 | - | 4 - 5 | 987 | 75 | |
| AK010520.1 | - | 3 - 4 | 883 | 75 | ||
| AK145941.1 | - | 4 - 5 | 864 | 75 | ||
| BC004029.1 | - | 4 - 5 | 873 | 3495093 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27880662 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2054455 | AK145941.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2054455 | NM_025510.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2054455 | AK009142.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2054456 | AK181511.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2054456 | BC004029.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2054456 | AK145941.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2054456 | NM_025510.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2054456 | AK009142.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 528400 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 883913 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 284480 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 54876 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 815079 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 722772 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 397423 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 365974 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 147408 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 136660 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P133718 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2054456 | AK181511.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2054456 | BC004029.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2054456 | AK145941.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2054456 | NM_025510.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2054456 | AK009142.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2054455 | AK145941.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2054455 | NM_025510.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2054455 | AK009142.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27880642 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27880640 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27880635 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27880636 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27880660 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27880650 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27880652 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27880659 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27880661 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27880633 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27880662 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27880644 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27880656 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27880646 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27880654 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27880639 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27880648 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P133718 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07378823 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07378800 | - | - |
| Whole Mouse Genome G4122A | A_52_P293724 | AK009142.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130841 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P133718 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09025280 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09025287 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27880662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880579 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880593 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880570 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27880602 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27880652 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27880606 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27880636 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27880639 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27880618 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27880658 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27880633 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27880627 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27880635 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09657857 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09307827 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27880611 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27880607 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27880644 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27880625 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09025275 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27880630 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09306165 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27880645 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27880659 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27880617 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27880623 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27880609 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27880612 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27880640 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09025273 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27880662 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27880620 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09458182 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27880660 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27880637 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27880654 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10387114 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387112 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387118 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387117 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387116 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387113 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1450987_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10387111 | - | - |
| Mouse Genome 430 2.0 Array | 1450987_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1450987_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66358_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66358_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100027193.mm.1 | NM_025510.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100027193.mm.1 | NM_025510.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100027193.mm.1 | TC1100002998.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100027196.mm.1 | NM_025510.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100027196.mm.1 | NM_025510.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100027196.mm.1 | TC1100002998.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100027194.mm.1 | NM_025510.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100027194.mm.1 | NM_025510.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100027194.mm.1 | TC1100002998.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100027195.mm.1 | NM_025510.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100027195.mm.1 | NM_025510.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100027195.mm.1 | TC1100002998.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1450987_PM_a_at | - | - |
| Mouse Expression Set 430 | 1450987_a_at | - | - |
| Murine Genome U74v2 Set | 104334_PM_at | - | - |
| Murine Genome U74v2 Set | 104334_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450987_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450987_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450987_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450987_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 753483 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 940397 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000940397 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1226476 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1226476 | - | - |
| mouse-6 v1.1 (Illumina) | scl40067.6.1_43-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 940397 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl40067.6.1_43-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 940397 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1226476 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1226476 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl40067.6.1_43-S | - | - |

Adprm
Genomic Map