Assay Details
Gene Symbol: Magohb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025564.2 | NP_079840.2 | 1 - 2 | 170 | 141 | |
GenBank mRNA | AK006303.1 | - | 1 - 2 | 160 | 141 | |
AK007769.1 | - | 1 - 2 | 157 | 141 | ||
AK008200.1 | - | 1 - 2 | 172 | 141 | ||
AK013310.1 | - | 1 - 2 | 155 | 141 | ||
BC027505.1 | - | 1 - 2 | 143 | 3372083 | 141 | |
KF051005.1 | - | 1 - 2 | 91 | 141 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24850240 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24850239 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24850238 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P187018 | AK013310.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P187018 | BC027505.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P187018 | AK007769.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P187018 | AK008200.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P187018 | NM_025564.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P187018 | AK013310.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P187018 | BC027505.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P187018 | AK007769.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P187018 | AK008200.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P187018 | NM_025564.2 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 153207 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 249602 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 403454 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 949001 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 404948 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 403531 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P175194 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P187018 | AK013310.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P187018 | BC027505.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P187018 | AK007769.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P187018 | AK008200.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P187018 | NM_025564.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P187018 | AK013310.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P187018 | BC027505.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P187018 | AK007769.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P187018 | AK008200.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P187018 | NM_025564.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24850279 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24850282 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24850253 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24850257 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24850264 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24850267 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24850229 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24850269 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24850236 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24850245 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24850281 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24850247 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24850240 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24850275 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24850239 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24850243 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24850277 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24850223 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24850268 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24850237 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24850230 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24850259 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24850272 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24850234 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24850241 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24850280 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24850244 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24850261 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24850250 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24850242 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24850265 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24850226 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24850254 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24850262 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24850271 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24850276 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24850225 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24850256 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24850232 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24850231 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24850274 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06070992 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P175194 | - | - |
Whole Mouse Genome G4122A | A_51_P187018 | AK013310.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P187018 | BC027505.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P187018 | AK007769.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P187018 | AK008200.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P187018 | NM_025564.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P187018 | AK013310.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P187018 | BC027505.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P187018 | AK007769.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P187018 | AK008200.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P187018 | NM_025564.2 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121729 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P175194 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05322162 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05322140 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05322160 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05818582 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05818579 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05322164 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24850238 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24850239 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24850240 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850197 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850280 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850145 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850271 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850245 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850232 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850265 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850176 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850223 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850220 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850199 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850204 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850217 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850257 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850214 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850188 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850243 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850229 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850272 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850236 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850259 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850149 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850158 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850190 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850254 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850166 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850244 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850153 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850192 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850137 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850131 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850247 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850256 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850151 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850141 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850173 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850239 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850219 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850284 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850241 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850203 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850234 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850185 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850200 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850183 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850136 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850143 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850193 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850206 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850283 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850138 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850178 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850264 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850163 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850207 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850139 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850250 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850267 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850184 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850226 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850147 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850180 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850189 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850261 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850167 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850182 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850282 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850152 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24850274 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24850210 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24850213 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05724701 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24850282 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24850229 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24850262 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24850265 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24850217 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24850279 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24850199 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05836214 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24850283 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24850203 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24850275 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24850239 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24850190 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24850192 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24850197 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24850250 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05817424 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05573071 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24850214 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24850220 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24850193 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24850254 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24850189 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05724697 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05818582 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24850261 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24850207 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05751619 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24850268 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24850188 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05706030 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24850204 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05322149 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05322140 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24850200 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05573069 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05322142 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24850271 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24850256 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24850257 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24850277 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24850274 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24850211 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24850226 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05724704 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24850206 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24850231 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24850244 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24850269 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24850280 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05322160 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24850267 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10548566 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10548570 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10548568 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10548572 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10548567 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10548569 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10548571 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1450943_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10548565 | - | - |
Mouse Genome 430 2.0 Array | 1450943_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1450943_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66441_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66441_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0600025697.mm.1 | TC0600003266.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600025697.mm.1 | NM_025564.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600025697.mm.1 | NM_025564.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600025701.mm.1 | TC0600003266.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600025701.mm.1 | NM_025564.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600025701.mm.1 | NM_025564.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600025704.mm.1 | TC0600003266.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600025704.mm.1 | NM_025564.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600025704.mm.1 | NM_025564.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600025695.mm.1 | TC0600003266.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600025695.mm.1 | NM_025564.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600025695.mm.1 | NM_025564.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600025703.mm.1 | TC0600003266.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600025703.mm.1 | NM_025564.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600025703.mm.1 | NM_025564.2 | Refseq |
Mouse Expression Set 430 | 1450943_PM_at | - | - |
Mouse Expression Set 430 | 1450943_at | - | - |
Mouse Genome 430A 2.0 Array | 1450943_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1450943_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1450943_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1450943_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 927157 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 510725 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0000510725 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1223176 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1223176 | - | - |
mouse-6 v1.1 (Illumina) | scl28314.7.1_50-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 510725 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl28314.7.1_50-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 510725 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1223176 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1223176 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl28314.7.1_50-S | - | - |
