Assay Details
| Gene Symbol: Ankrd61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025732.3 | NP_080008.1 | 1 - 2 | 297 | 63 | |
| GenBank mRNA | AK014929.1 | - | 1 - 2 | 298 | 63 | |
| AK017053.1 | - | 1 - 2 | 238 | 63 | ||
More Information
Set Membership: |
|

Ankrd61
Genomic Map