Assay Details
Gene Symbol: Acot13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025790.2 | NP_080066.1 | 1 - 2 | 268 | 101 | |
GenBank mRNA | AK002261.1 | - | 1 - 2 | 270 | 101 | |
AK008624.1 | - | 1 - 2 | 226 | 101 | ||
AK144523.1 | - | 1 - 2 | 272 | 101 | ||
AK212217.1 | - | 1 - 2 | 101 | |||
BC096567.1 | - | 1 - 2 | 267 | 1314880 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
metabolic process
protein homotetramerization
CoA hydrolase activity
hydrolase activity
2-oxoglutaryl-CoA thioesterase activity
2,4,4-trimethyl-3-oxopentanoyl-CoA thioesterase activity
3-isopropylbut-3-enoyl-CoA thioesterase activity
glutaryl-CoA hydrolase activity
acyl-CoA hydrolase activity
metabolic process
protein homotetramerization
CoA hydrolase activity
hydrolase activity
2-oxoglutaryl-CoA thioesterase activity
2,4,4-trimethyl-3-oxopentanoyl-CoA thioesterase activity
3-isopropylbut-3-enoyl-CoA thioesterase activity
glutaryl-CoA hydrolase activity
acyl-CoA hydrolase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28889205 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28889206 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28889207 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P502422 | AK002261.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P502422 | AK144523.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P502422 | BC018165.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P502422 | BC096567.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P502422 | NM_025790.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P502422 | AK002261.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P502422 | AK144523.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P502422 | BC018165.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P502422 | BC096567.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P502422 | NM_025790.2 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 508259 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 743622 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 265222 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 481058 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 953294 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 837518 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 305222 | AK019729.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 631451 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 929053 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 699943 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 83360 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 153066 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P135608 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P135410 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P502422 | AK002261.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P502422 | AK144523.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P502422 | BC018165.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P502422 | BC096567.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P502422 | NM_025790.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P502422 | AK002261.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P502422 | AK144523.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P502422 | BC018165.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P502422 | BC096567.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P502422 | NM_025790.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28889205 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28889183 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28889196 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28889203 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28889204 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28889201 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28889180 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28889185 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28889184 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28889198 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28889189 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28889206 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28889187 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28889197 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28889192 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28889200 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28889179 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28889207 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28889199 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28889195 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P135410 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P135608 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07813168 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07813184 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07813160 | - | - |
Whole Mouse Genome G4122A | A_51_P502422 | AK002261.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P502422 | AK144523.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P502422 | BC018165.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P502422 | BC096567.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P502422 | NM_025790.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P502422 | AK002261.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P502422 | AK144523.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P502422 | BC018165.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P502422 | BC096567.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P502422 | NM_025790.2 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P105906 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136528 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P135608 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P135410 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10572164 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10572163 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10572159 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10572167 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28889207 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28889205 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28889206 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28889090 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28889078 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28889187 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28889113 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28889163 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28889104 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28889168 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28889126 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28889198 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28889117 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28889087 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28889170 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28889204 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28889116 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28889096 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28889110 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28889199 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28889083 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28889136 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28889167 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28889174 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28889101 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28889192 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28889145 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28889081 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28889207 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28889080 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28889205 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28889176 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28889074 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28889139 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28889132 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28889203 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28889111 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28889129 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28889185 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28889119 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28889180 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28889164 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28889171 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28889189 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28889076 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28889073 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28889201 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28889183 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28889085 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28889123 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28889177 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28889196 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28889128 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28889160 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28889140 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28889151 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28889106 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28889077 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28889155 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28889131 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28889166 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28889144 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28889091 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28889169 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28889138 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28889204 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28889195 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10627348 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28889179 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10572168 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28889191 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28889190 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28889171 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10512443 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28889183 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10554952 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28889185 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28889192 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28889196 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28889201 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28889205 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28889198 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11025056 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11025055 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28889203 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10834662 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28889189 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28889199 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10566866 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28889197 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28889187 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28889166 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10731109 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10408332 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10408333 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10408334 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1417316_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10408331 | - | - |
Mouse Genome 430 2.0 Array | 1417316_at | - | - |
Mouse Genome 430 2.0 Array | 1417316_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66834_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66834_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1300010384.mm.1 | NM_025790.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300010384.mm.1 | NM_025790.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300010384.mm.1 | TC1300001669.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1300010383.mm.1 | NM_025790.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300010383.mm.1 | NM_025790.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300010383.mm.1 | TC1300001669.mm.1 | MTA TC |
Mouse Expression Set 430 | 1417316_at | - | - |
Mouse Expression Set 430 | 1417316_PM_at | - | - |
Murine Genome U74v2 Set | 93780_PM_at | - | - |
Murine Genome U74v2 Set | 93780_at | - | - |
Mouse Genome 430A 2.0 Array | 1417316_at | - | - |
Mouse Genome 430A 2.0 Array | 1417316_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1417316_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1417316_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 401451 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 6840746 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006840746 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2604338 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2604338 | - | - |
mouse-6 v1.1 (Illumina) | scl44185.3.1_24-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6840746 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl44185.3.1_24-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6840746 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2961626 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2961626 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl44185.3.1_24-S | - | - |
