Assay Details
Gene Symbol: Fuca2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025799.4 | NP_080075.2 | 3 - 4 | 879 | 74 | |
GenBank mRNA | AK002675.1 | - | 3 - 4 | 821 | 74 | |
AK008653.1 | - | 3 - 4 | 824 | 74 | ||
AK017443.1 | - | 3 - 4 | 849 | 74 | ||
AK030320.1 | - | 3 - 4 | 881 | 74 | ||
AK035607.1 | - | 3 - 4 | 760 | 74 | ||
AK044790.1 | - | 3 - 4 | 736 | 74 | ||
AK146295.1 | - | 3 - 4 | 808 | 74 | ||
AK151204.1 | - | 3 - 4 | 740 | 74 | ||
AK159288.1 | - | 3 - 4 | 825 | 74 | ||
BC004039.1 | - | 3 - 4 | 830 | 3496187 | 74 | |
BC018181.1 | - | 3 - 4 | 839 | 3983067 | 74 | |
BC094449.1 | - | 3 - 4 | 794 | 4222249 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
