Assay Details
| Gene Symbol: Fuca2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025799.4 | NP_080075.2 | 6 - 7 | 1387 | 73 | |
| GenBank mRNA | AK002675.1 | - | 6 - 7 | 1329 | 73 | |
| AK008653.1 | - | 6 - 7 | 1332 | 73 | ||
| AK017443.1 | - | 6 - 7 | 1357 | 73 | ||
| AK030320.1 | - | 6 - 7 | 1389 | 73 | ||
| AK035607.1 | - | 6 - 7 | 1268 | 73 | ||
| AK044790.1 | - | 6 - 7 | 1244 | 73 | ||
| AK146295.1 | - | 6 - 7 | 1316 | 73 | ||
| AK151204.1 | - | 6 - 7 | 1248 | 73 | ||
| AK159288.1 | - | 6 - 7 | 1333 | 73 | ||
| AK205091.1 | - | 1 - 2 | 121 | 73 | ||
| AK218452.1 | - | 1 - 2 | 121 | 73 | ||
| BC004039.1 | - | 6 - 7 | 1338 | 3496187 | 73 | |
| BC018181.1 | - | 6 - 7 | 1347 | 3983067 | 73 | |
| BC094449.1 | - | 6 - 7 | 1302 | 4222249 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P03719 | BC004039.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P03719 | BC018181.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P03719 | AK159288.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P03719 | AK205091.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P03719 | AK218452.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2102529 | BC004039.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2102529 | BC018181.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2102529 | BC094449.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2102529 | AK159288.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2102529 | AK044790.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2102533 | AK030320.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2102533 | NM_025799.4 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 434382 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 139884 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 98540 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 593896 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 50981 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 816473 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 703048 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 969219 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 173188 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 22729 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P127617 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P117622 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2102533 | AK030320.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2102533 | NM_025799.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2102529 | BC004039.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2102529 | BC018181.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2102529 | BC094449.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2102529 | AK159288.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2102529 | AK044790.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P436957 | AK159288.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P436957 | AK044790.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P436957 | AK030320.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P436957 | AK035607.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P436957 | NM_025799.4 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26951528 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26951524 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26951544 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26951516 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26951521 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26951537 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26951515 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26951546 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26951543 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26951533 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26951512 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26951549 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26951518 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26951525 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26951520 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26951527 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26951534 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26951532 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26951539 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26951531 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26951535 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26951522 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P117622 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06980564 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06980586 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P127617 | - | - |
| Whole Mouse Genome G4122A | A_51_P436957 | AK159288.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P436957 | AK044790.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P436957 | AK030320.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P436957 | AK035607.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P436957 | NM_025799.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P441949 | AK030320.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P441949 | NM_025799.4 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136070 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P117622 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P127617 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08400582 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08452337 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08652523 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08400578 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08400579 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26951520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26951567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26951616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26951571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26951626 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26951548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26951622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26951496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26951623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26951543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26951599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26951533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26951546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26951531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26951593 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26951532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26951507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26951632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26951524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26951508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26951560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26951554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26951502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26951595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26951527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26951608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26951617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26951577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26951518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26951525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26951564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26951631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26951559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26951515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26951522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26951556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26951503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26951539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26951597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26951544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26951566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26951512 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26951583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26951561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26951601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26951568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26951614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26951573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26951587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26951565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26951500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26951627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26951610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26951498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26951574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26951607 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26951619 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26951571 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26951559 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08759376 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26951561 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26951607 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26951617 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26951557 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26951568 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08466567 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26951564 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26951533 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26951532 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26951512 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26951544 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26951554 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26951556 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26951546 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08652522 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26951614 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08400583 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08306640 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26951528 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08805355 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26951610 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26951531 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26951573 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26951558 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26951574 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26951520 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26951527 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26951518 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26951552 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26951543 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26951566 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26951577 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08805358 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08306636 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26951509 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26951539 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26951522 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26951565 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08833830 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26951537 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26951535 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26951541 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26951529 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26951510 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10361794 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10361798 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10361796 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10361795 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10361797 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10361792 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10361791 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10361793 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1440313_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416047_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416046_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10361790 | - | - |
| Mouse Genome 430 2.0 Array | 1416046_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1440313_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1416047_at | - | - |
| Mouse Genome 430 2.0 Array | 1440313_at | - | - |
| Mouse Genome 430 2.0 Array | 1416047_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1416046_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66848_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66848_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1000000714.mm.1 | NM_025799.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000000714.mm.1 | TC1000000095.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000000721.mm.1 | NM_025799.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000000721.mm.1 | TC1000000095.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000000712.mm.1 | NM_025799.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000000712.mm.1 | TC1000000095.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000000713.mm.1 | NM_025799.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000000713.mm.1 | TC1000000095.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000000715.mm.1 | NM_025799.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000000715.mm.1 | TC1000000095.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000000720.mm.1 | NM_025799.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000000720.mm.1 | TC1000000095.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1416046_PM_a_at | - | - |
| Mouse Expression Set 430 | 1416047_PM_at | - | - |
| Mouse Expression Set 430 | 1416047_at | - | - |
| Mouse Expression Set 430 | 1416046_a_at | - | - |
| Mouse Expression Set 430 | 1440313_PM_at | - | - |
| Mouse Expression Set 430 | 1440313_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416046_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416047_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416046_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416047_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416046_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416047_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416047_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416046_a_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440313_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440313_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 3360739 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003360739 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2621780 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2621780 | - | - |
| mouse-6 v1.1 (Illumina) | scl39142.8_281-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3360739 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl39142.8_281-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3360739 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2621780 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2621780 | - | - |

Fuca2
Genomic Map