Assay Details
| Gene Symbol: Pcyox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025823.4 | NP_080099.1 | 5 - 6 | 926 | 90 | |
| GenBank mRNA | AK004799.2 | - | 5 - 6 | 881 | 90 | |
| AK004840.1 | - | 5 - 6 | 881 | 90 | ||
| AK031585.1 | - | 5 - 6 | 882 | 90 | ||
| AK033373.1 | - | 5 - 6 | 882 | 90 | ||
| AK049412.1 | - | 5 - 6 | 879 | 90 | ||
| AK134528.1 | - | 5 - 6 | 927 | 90 | ||
| AK145366.1 | - | 5 - 6 | 886 | 90 | ||
| AK147190.1 | - | 5 - 6 | 894 | 90 | ||
| AK159981.1 | - | 5 - 6 | 882 | 90 | ||
| AK161616.1 | - | 5 - 6 | 882 | 90 | ||
| AK169209.1 | - | 5 - 6 | 882 | 90 | ||
| AK173058.1 | - | 5 - 6 | 848 | 90 | ||
| BC006056.1 | - | 3 - 4 | 537 | 90 | ||
| BC028308.1 | - | 5 - 6 | 896 | 3486530 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
chloride transport
prenylated protein catabolic process
prenylcysteine catabolic process
oxidation-reduction process
chloride transmembrane transport
prenylcysteine oxidase activity
chloride-transporting ATPase activity
oxidoreductase activity
oxidoreductase activity, acting on a sulfur group of donors, oxygen as acceptor
chloride transport
prenylated protein catabolic process
prenylcysteine catabolic process
oxidation-reduction process
chloride transmembrane transport
prenylcysteine oxidase activity
chloride-transporting ATPase activity
oxidoreductase activity
oxidoreductase activity, acting on a sulfur group of donors, oxygen as acceptor
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24609840 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24609841 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24609842 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24609843 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P508029 | BC028308.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P508029 | AK173058.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P508029 | AK161616.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P508029 | AK147190.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P508029 | AK134528.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P508029 | BC028308.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P508029 | AK173058.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P508029 | AK161616.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P508029 | AK147190.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P508029 | AK134528.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2156293 | BC006056.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2156293 | BC028308.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2156293 | AK173058.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2156293 | AK161616.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2156293 | AK147190.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 158432 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 486672 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 944885 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 893398 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 800853 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 407769 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 456308 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 155724 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 48130 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P508029 | BC028308.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P508029 | AK173058.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P508029 | AK161616.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P508029 | AK147190.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P508029 | AK134528.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P508029 | BC028308.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P508029 | AK173058.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P508029 | AK161616.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P508029 | AK147190.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P508029 | AK134528.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2156293 | BC006056.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2156293 | BC028308.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2156293 | AK173058.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2156293 | AK161616.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2156293 | AK147190.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P508029 | BC028308.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P508029 | AK173058.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P508029 | AK161616.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P508029 | AK147190.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P508029 | AK134528.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P508029 | BC028308.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P508029 | AK173058.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P508029 | AK161616.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P508029 | AK147190.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P508029 | AK134528.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05968804 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01375413 | - | - |
| Whole Mouse Genome G4122A | A_51_P508029 | BC028308.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P508029 | AK173058.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P508029 | AK161616.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P508029 | AK147190.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P508029 | AK134528.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P508029 | BC028308.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P508029 | AK173058.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P508029 | AK161616.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P508029 | AK147190.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P508029 | AK134528.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P133682 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05968804 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05235686 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05235702 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05235688 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05235689 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24609843 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24609840 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24609841 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24609842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24609812 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05294626 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24609850 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24609849 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24609855 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24609848 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05294623 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24609856 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10545914 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10545911 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10545915 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10545912 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10545913 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10545916 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1446325_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1423394_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1447277_PM_s_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10545910 | - | - |
| Mouse Genome 430 2.0 Array | 1447277_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1423394_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1423394_at | - | - |
| Mouse Genome 430 2.0 Array | 1446325_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1441834_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1447277_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1446325_at | - | - |
| Mouse Genome 430 2.0 Array | 1441834_PM_x_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66881_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66881_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600020448.mm.1 | TC0600002635.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600020448.mm.1 | NM_025823.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600020448.mm.1 | NM_025823.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600020450.mm.1 | TC0600002635.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600020450.mm.1 | NM_025823.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600020450.mm.1 | NM_025823.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600020449.mm.1 | TC0600002635.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600020449.mm.1 | NM_025823.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600020449.mm.1 | NM_025823.4 | Refseq |
| Mouse Expression Set 430 | 1423394_PM_at | - | - |
| Mouse Expression Set 430 | 1423394_at | - | - |
| Mouse Expression Set 430 | 1446325_PM_at | - | - |
| Mouse Expression Set 430 | 1446325_at | - | - |
| Mouse Expression Set 430 | 1447277_PM_s_at | - | - |
| Mouse Expression Set 430 | 1447277_s_at | - | - |
| Murine Genome U74v2 Set | 97485_at | - | - |
| Murine Genome U74v2 Set | 97485_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423394_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423394_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423394_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423394_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1446325_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1447277_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1446325_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1447277_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 632833 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 870161 | - | - |
| MouseWG-6 v2 Expression BeadChip | 105720575 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 105720575 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 870161 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000870161 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1258607 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1258607 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2472504 | - | - |
| mouse-6 v1.1 (Illumina) | scl28753.6_75-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0066881.2_144-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 870161 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 105720575 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0066881.2_144-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 870161 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 105720575 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2472504 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2472504 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0066881.2_144-S | - | - |

Pcyox1
Genomic Map