Assay Details
Gene Symbol: Proz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025834.3 | NP_080110.1 | 6 - 7 | 710 | 72 | |
GenBank mRNA | AK005011.2 | - | 6 - 7 | 654 | 72 | |
AK008819.1 | - | 6 - 7 | 711 | 72 | ||
BC024104.1 | - | 6 - 7 | 839 | 5136392 | 72 | |
BC038045.1 | - | 6 - 7 | 675 | 4196827 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P170959 | AK005011.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P170959 | AK005011.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P356068 | BC024104.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P356068 | AK005011.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P356068 | AK008819.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P356068 | NM_025834.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P356068 | BC024104.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P356068 | AK005011.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P356068 | AK008819.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P356068 | NM_025834.3 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 257868 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 896438 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 145606 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 669189 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 753612 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 824537 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 925290 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 713759 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 491775 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 119615 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 4948 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 673822 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 120488 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P176582 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P356068 | BC024104.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P356068 | AK005011.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P356068 | AK008819.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P356068 | NM_025834.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P356068 | BC024104.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P356068 | AK005011.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P356068 | AK008819.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P356068 | NM_025834.3 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P203398 | BC024104.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P203398 | BC038045.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P203398 | AK005011.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P203398 | AK008819.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P203398 | NM_025834.3 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P170959 | AK005011.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P170959 | AK005011.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25665862 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25665880 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25665853 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25665891 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25665846 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25665877 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25665866 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25665852 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25665855 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25665885 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25665890 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25665888 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25665854 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25665868 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25665873 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25665858 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25665870 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25665857 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25665882 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25665864 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25665861 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25665860 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25665875 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25665886 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25665881 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25665849 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P176582 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06428782 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01677908 | - | - |
Whole Mouse Genome G4122A | A_52_P356068 | BC024104.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P356068 | AK005011.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P356068 | AK008819.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P356068 | NM_025834.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P356068 | BC024104.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P356068 | AK005011.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P356068 | AK008819.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P356068 | NM_025834.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P203398 | BC024104.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P203398 | BC038045.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P203398 | AK005011.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P203398 | AK008819.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P203398 | NM_025834.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P170959 | AK005011.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P170959 | AK005011.2 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P111461 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P176582 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07091642 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07091632 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07091636 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07091635 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07091638 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25665927 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25665916 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25665862 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25665896 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25665854 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25665963 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25665864 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25665967 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25665894 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25665857 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25665906 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25665971 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25665883 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25665849 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25665868 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25666033 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25665882 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25665907 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25665964 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25666036 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25665861 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25665902 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25665875 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25665890 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25666034 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25665917 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25665893 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25665933 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25665853 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25665914 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25665931 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25665965 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25665899 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25665900 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25665892 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25665909 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25665918 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25665957 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25665870 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25665920 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25665934 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25665962 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25665878 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25665888 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25665904 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25665873 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25665960 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25665979 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25665866 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25665898 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25665958 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25665887 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25665855 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25665885 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25665860 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25665852 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07379020 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25665882 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25665853 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07379023 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25665892 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25665887 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07410565 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25665883 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07410555 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25665900 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25665888 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25665877 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06992578 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25665855 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07410564 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25665854 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07018274 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25665868 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07305858 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25665849 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25665911 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06722915 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25665860 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25665873 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25665902 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25665929 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07448048 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25665864 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25665870 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25665852 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25665917 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25665893 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25665906 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07123430 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25665878 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25665904 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25665914 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25665875 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25665899 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25665890 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25665918 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25665920 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25665885 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25665925 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25665898 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07410579 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25665909 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25665912 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25665896 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10570318 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10570314 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10570316 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10570309 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10570319 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10570315 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10570312 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10570310 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10570311 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10570313 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10570317 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1450201_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1431721_PM_a_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10570308 | - | - |
Mouse Genome 430 2.0 Array | 1450201_at | - | - |
Mouse Genome 430 2.0 Array | 1431721_a_at | - | - |
Mouse Genome 430 2.0 Array | 1431721_PM_a_at | - | - |
Mouse Genome 430 2.0 Array | 1450201_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66901_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66901_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0800001085.mm.1 | NM_025834.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800001085.mm.1 | NM_025834.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800001085.mm.1 | TC0800000117.mm.1 | MTA TC |
Mouse Expression Set 430 | 1450201_PM_at | - | - |
Mouse Expression Set 430 | 1431721_a_at | - | - |
Mouse Expression Set 430 | 1431721_PM_a_at | - | - |
Mouse Expression Set 430 | 1450201_at | - | - |
Mouse Genome 430A 2.0 Array | 1450201_at | - | - |
Mouse Genome 430A 2.0 Array | 1450201_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1431721_PM_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1431721_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1450201_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1431721_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1431721_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1450201_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 537369 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 105390152 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 3440541 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0003440541 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2736621 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2736621 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2497831 | - | - |
mouse-6 v1.1 (Illumina) | scl0066901.2_72-S | - | - |
mouse-6 v1.1 (Illumina) | scl34061.9_165-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 3440541 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 105390152 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0066901.2_72-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 105390152 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3440541 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2736621 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2497831 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2736621 | - | - |
