Assay Details
| Gene Symbol: Acad8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025862.2 | NP_080138.2 | 3 - 4 | 429 | 76 | |
| GenBank mRNA | AK009379.1 | - | 3 - 4 | 408 | 76 | |
| AK019502.1 | - | 3 - 4 | 426 | 76 | ||
| AK029897.1 | - | 3 - 4 | 393 | 76 | ||
| AK051091.1 | - | 3 - 4 | 384 | 76 | ||
| AK077335.1 | - | 3 - 4 | 409 | 76 | ||
| AK161255.1 | - | 3 - 4 | 383 | 76 | ||
| AK211554.1 | - | 1 - 2 | 148 | 76 | ||
| BC026535.1 | - | 3 - 4 | 403 | 76 | ||
| BC037644.1 | - | 3 - 4 | 405 | 5053565 | 76 | |
| BQ931973.1 | - | 3 - 4 | 429 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Acad8
Genomic Map