Assay Details
| Gene Symbol: Acad8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025862.2 | NP_080138.2 | 8 - 9 | 986 | 61 | |
| GenBank mRNA | AK009379.1 | - | 8 - 9 | 965 | 61 | |
| AK019502.1 | - | 8 - 9 | 983 | 61 | ||
| AK029897.1 | - | 8 - 9 | 950 | 61 | ||
| AK049908.1 | - | 4 - 5 | 1302 | 61 | ||
| AK077335.1 | - | 8 - 9 | 966 | 61 | ||
| AK161255.1 | - | 8 - 9 | 940 | 61 | ||
| BC026535.1 | - | 8 - 9 | 960 | 61 | ||
| BC037644.1 | - | 8 - 9 | 962 | 5053565 | 61 | |
More Information
Set Membership: |
|

Acad8
Genomic Map