Assay Details
| Gene Symbol: Acad8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025862.2 | NP_080138.2 | 9 - 10 | 1135 | 65 | |
| GenBank mRNA | AK009379.1 | - | 9 - 10 | 1114 | 65 | |
| AK019502.1 | - | 9 - 10 | 1132 | 65 | ||
| AK029897.1 | - | 9 - 10 | 1099 | 65 | ||
| AK051091.1 | - | 9 - 10 | 1090 | 65 | ||
| AK077335.1 | - | 9 - 10 | 1115 | 65 | ||
| BC026535.1 | - | 9 - 10 | 1109 | 65 | ||
| BC037644.1 | - | 9 - 10 | 1111 | 5053565 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Acad8
Genomic Map